Can the HulaMixer sample mixer be used at low temperature?
The HulaMixer sample mixer may be used at 4 degrees C but not for an extended period of time. The high humidity and low temperature could eventually lead to rust and reduce the lifetime of the mixer.