Pierce™ Firefly Luciferase Flash Assay Kit
Thermo Scientific™

Pierce™ Firefly Luciferase Flash Assay Kit

Assay regulatory element transcriptional activity in whole cell lysates with this highly sensitive flash-based firefly luciferase kit.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
161751000 Reaction Kit
16174100 Reaction Kit
Catalog number 16175
Price (USD)
716.00
Each
Quantity:
1000 Reaction Kit
Price (USD)
716.00
Each

The Thermo Scientific Pierce Firefly Luciferase Flash Assay Kit provides reagents to measure Red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.

Features of the Firefly Luciferase Flash Assay Kit:

  • Highly sensitive detection of firefly luciferase activity
  • Use with Pierce Firefly Luciferase Signal Enhancer (Cat. No. 16180) for best results (maximum signal)
  • Working solution stable for up to 2 months at -20°C
  • Assay reagents compatible with other firefly luciferases
  • Amenable to automation
  • Contains a universal cell lysis buffer and optimized flash assay reagent
  • Safe non-radioactive assay

Thermo Scientific Pierce Firefly Luciferase Flash Assay Kit contains reagents to measure the activity of firefly luciferase in mammalian whole cell lysates. Containing a convenient lysis buffer, assay buffer and firefly substrate, this kit provides a complete reagent system for the intracellular detection of promoter or pathway activity. Light output resulting from the luciferase reaction is captured using a luminometer and can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression. These reagents are optimized to work with Thermo Scientific Firefly Luc Plasmids; however, the Flash Assay Kit may be used to detect the activity of other firefly or ATP-dependent luciferases utilizing D-luciferin as a substrate.

Includes:

Cell lysis buffer, reaction buffer and substrate; purchase Luciferase Signal Enhancer (Cat. No. 16180) separately.

Requires:

  • Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers.
  • For best results (maximum signal), use with Pierce Firefly Luciferase Signal Enhancer (Cat. No. 16180)

Applications:

  • Promoter studies for analyzing cis- regulatory elements and trans-acting factors
  • Drug screening
  • siRNA and miRNA screening
  • Multiplexed assays to study off-target effects
  • Secretory pathway/protein localization reporter assays
  • Signal transduction pathway analysis
  • RNA splicing studies

Firefly luciferase is a 60kDa protein produced in nature by several species of the Lampyridae family of beetles which includes the genera Photinus and Luciola. Bioluminescent signal from firefly luciferase originates from the oxidation of D-luciferin. Light output captured using a luminometer can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression.

For Research Use Only. Not for use in diagnostic procedures.
Firefly luciferase is a 60kDa protein produced in nature by several species of the Lampyridae family of beetles which includes the genera Photinus and Luciola. Bioluminescent signal from firefly luciferase originates from the oxidation of D-luciferin. Light output captured using a luminometer can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression.
Specifications
AssayReporter Enzyme, Luciferase Reporter Assay
Compatible CellsMammalian Cells
Detection MethodBioluminescence
For Use With (Equipment)Luminometer
Format96-well Plate, 384-well Plate
Label TypeEnzyme Labeled
Product LinePierce
Quantity1000 Reaction Kit
SubstrateD-Luciferin
Substrate PropertiesChemical Substrate
Substrate TypeLuciferase Substrate
TargetLuciferase, Firefly Luciferase
TechniqueEnhanced Chemiluminescence
TypeAssay Kit
Unit SizeEach
Contents & Storage
• Firefly Flash Assay Buffer, 5 mL, store at 4°C
• D-Luciferin, 3 mg, store at 4°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature

Upon receipt store kit at 4°C or store individual components as indicated above.

Frequently asked questions (FAQs)

I got a high background signal with a Firefly Luciferase Glow/Flash assay. Why is this?

This could be due to contamination of the control sample. Make sure to use new sample and change pipette tips after each well.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I got a high signal with a Firefly Luciferase Glow/Flash assay. What could have happened?

This could be due to high luciferase expression. Here are some suggestions:

  • Reduce incubation time before collecting samples. 
  • Decrease the integration time on the instrument.
  • Dilute the sample 
    Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay



  • Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

    What could be causing no signal or low signal with the Firefly Luciferase Glow/Luciferase Flash assay? Do you have any suggestions to get around this problem?

    Here are possible causes and solutions:

    - Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality - use only transfection grade DNA; Use actively dividing, low passage cells; Use a different cell type.
    - No or low promoter activity: Use conditions known for promoter activation; Incubate cells for a longer time; Change growth conditions to improve expression; Use a different promoter.
    - D-luciferin auto-oxidized: Protect substrate from light and maintain 100X D-luciferin at -20 degrees C; Prepare new Working Solution if used longer than 4 hours.
    - Low luciferase expression: Use Pierce Firefly Signal Enhancer (100X) (Cat. No. 16180); Lyse cells in a smaller volume of 1X Cell Lysis Buffer; Use a different promoter or growth conditions to improve expression; Increase the integration time on the instrument; Scale-up the volume of sample and reagent per well.
    - Degraded luciferase protein: Store cell lysates on ice and perform assays immediately following cell lysis.

    Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

    I got a high background signal with a Gaussia/Cypridina/Renilla Luciferase Glow/Luciferase Flash assay. What went wrong?

    Here are possible causes and solutions:

    - Non-specific oxidation of substrate: Use less serum in the cell culture media; Note: Albumin can increase the auto-oxidation of Vargulin; Avoid repeated freezing and thawing of the sample.
    - Control sample is contaminated: Use new sample; Change pipette tips after each well.

    Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

    I am getting a high saturating signal with a Gaussia/Cypridina/Renilla Luciferase Glow/Luciferase Flash assay. What could have happened?

    This could be due to high luciferase expression. Here are some suggestions:

  • Reduce incubation time before collecting samples. 
  • Decrease the integration time on the instrument.
  • Dilute the sample: for secreted Gaussia/Cypridina Luciferase, dilute the sample using media from the cell culture; for cell lysate, dilute the sample using lysis buffer 


  • Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.

    Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.