Thermo Scientific Pierce Firefly Signal Enhancer is designed to complement the Pierce Firefly Luciferase Flash and Glow Assay and isRead more
Have Questions?
Catalog Number
Quantity
16180
0.5 mL
Catalog number 16180
Price (USD)
44.49
Special offer
Online exclusive
Ends: 31-Dec-2026
54.25
Save 9.76
Each
Quantity:
0.5 mL
Price (USD)
44.49
Special offer
Online exclusive
Ends: 31-Dec-2026
54.25
Save 9.76
Each
Thermo Scientific Pierce Firefly Signal Enhancer is designed to complement the Pierce Firefly Luciferase Flash and Glow Assay and is recommended when maximum sensitivity is desired.
Features of Firefly Signal Enhancer:
• Up to 5-fold increase in Firefly Flash signal intensity • Up to 2-fold increase in Firefly Glow signal intensity and duration • Convenient and easy-to-use 100X solution
Pierce Firefly Signal Enhancer is designed to to be used with the Pierce Firefly Luciferase Flash Assay Kit (Part No. 16174, 16175) and Pierce Firefly Luciferase Glow Assay Kit (Part No. 16176, 16177). When used with the flash assay kit, the enhancer provides a five-fold increase in signal. When used with the glow assay kit, the enhancer solution increases the signal strength and duration up to two-fold and also increases stability of the bioluminescent signal.
Requires: Pierce Firefly Luciferase Flash (Part No. 16174, 16175) or Glow (Part No. 16176, 16177) Assay Kits; Luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodBioluminescence
Product LinePierce
Quantity0.5 mL
Substrate PropertiesChemical Substrate
Substrate TypeLuciferase Substrate
TypeFirefly Signal Enhancer
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.