Goat Serum, New Zealand origin, 500 mL - FAQs
Goat Serum, New Zealand origin, 500 mL - FAQs
View additional product information for Goat Serum, New Zealand origin - FAQs (16210064, 16210072)
2 product FAQs found
Does Goat Serum, New Zealand origin (Cat. Nos. 16210064, 16210072) need to be heat-inactivated before use, or is it ready for use as is? If it does need to be heat-inactivated, what is the protocol to do so?
Goat Serum, New Zealand origin is not heat-inactivated. You can heat-inactivate by incubating the fully thawed, unopened serum bottle in a 56 degrees C water bath for 30 min. Be sure to swirl the bottle several times during the 30 min incubation.