Adonitol, 98%
Adonitol, 98%
Adonitol, 98%
Thermo Scientific Chemicals

Adonitol, 98%

Adonitol, 98%, C5H12O5, CAS Number-488-81-3, adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol, 100g, 97.5% min. (ELSD) (HPLC), 01,53, 100.0, 99.0, White, 207-685-7, 152.15, HEBKCHPVOIAQTA-NGQZWQHPSA-N, Authentic, (2S,4R)-pentane-1,2,3,4,5-pentol | CAS: 488-81-3 | C5H12O5 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 167035000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS488-81-3
IUPAC Namepentane-1,2,3,4,5-pentol
Molecular FormulaC5H12O5
InChI KeyHEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Infrared spectrumConforms
Melting point101°C to 105°C
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only