Nunc™ Cell Culture/Petri Dishes, 145 cm2, Nunclon Delta treated, with Airvent, Pack of 10 - FAQs
Nunc™ Cell Culture/Petri Dishes, 145 cm2, Nunclon Delta treated, with Airvent, Pack of 10 - FAQs
View additional product information for Nunc™ Cell Culture/Petri Dishes - FAQs (153066, 168381, 150318, 174926, 150326, 169558, 174888, 172931, 150288, 150350, 171099)
2 product FAQs found
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?
Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.
The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells
More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .
What is Nunclon Delta?
Nunclon Delta refers to our performance-certified surface for culture of adherent cells, available in a variety of common formats (flasks, plates, multiwell dishes). It is tested with at least four cell lines (varies by product and manufacturing location) commonly used in research, and it should work well for most robust cell lines. It does not involve addition of coating reagent, but rather is manufactured by exposure of the molded products to a proprietary energy source, which renders the surface more hydrophilic by means of increasing the concentration of oxygen-containing (e.g. hydroxyl) molecular groups. It is by this hydrophilic property that anchorage-dependent cells are able to attach to the surface and become confluent.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.