Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Thermo Scientific™

Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes

Increase filtration efficiency. PES membranes and exclusive Rapid-Flow design provide fast flow rates and high throughput while defending against cell culture contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeMaterial (Membrane)Diameter (Metric) MembraneVolume (Metric) ReceiverUnit Size
169-00450.45 μmPES90 mm500 mLCase of 12
564-00200.2 μmPES50 mm50 mLCase of 12
124-00450.45 μmPES50 mm115 mLCase of 72
124-0045PK0.45 μmPES50 mm115 mLPack of 12
565-00100.1 μmPES50 mm150 mLCase of 12
524-00200.2 μmPES50 mm115 mLCase of 72
565-00200.2 μmPES50 mm150 mLCase of 12
165-00450.45 μmPES50 mm150 mLCase of 12
568-00100.1 μmPES75 mm250 mLCase of 12
568-00200.2 μmPES50 mm250 mLCase of 12
168-00450.45 μmPES50 mm250 mLCase of 12
566-00100.1 μmPES75 mm500 mLCase of 12
566-00200.2 μmPES75 mm500 mLCase of 12
569-00200.2 μmPES90 mm500 mLCase of 12
166-00450.45 μmPES75 mm500 mLCase of 12
567-00100.1 μmPES90 mm1000 mLCase of 12
567-00200.2 μmPES90 mm1000 mLCase of 12
167-00450.45 μmPES90 mm1000 mLCase of 12
121-00200.2 μmCellulose Nitrate50 mm115 mLCase of 72
121-0020PK
also known as 121-0020
0.2 μmCellulose Nitrate50 mm115 mLPack of 12
121-00450.45 μmCellulose Nitrate50 mm115 mLCase of 72
121-0045PK0.45 μmCellulose Nitrate50 mm115 mLPack of 12
125-00200.2 μmCellulose Nitrate50 mm150 mLCase of 12
125-00450.45 μmCellulose Nitrate50 mm150 mLCase of 12
125-00800.8 μmCellulose Nitrate50 mm150 mLCase of 12
126-00200.2 μmCellulose Nitrate50 mm250 mLCase of 12
126-00450.45 μmCellulose Nitrate50 mm250 mLCase of 12
126-00800.8 μmCellulose Nitrate50 mm250 mLCase of 12
450-00200.2 μmCellulose Nitrate75 mm500 mLCase of 12
450-00450.45 μmCellulose Nitrate75 mm500 mLCase of 12
450-00800.8 μmCellulose Nitrate75 mm500 mLCase of 12
127-00200.2 μmCellulose Nitrate75 mm1000 mLCase of 12
127-00450.45 μmCellulose Nitrate75 mm1000 mLCase of 12
127-00800.8 μmCellulose Nitrate75 mm1000 mLCase of 12
122-00200.2 μmSFCA50 mm115 mLCase of 72
122-0020PK
also known as 122-0020
0.2 μmSFCA50 mm115 mLPack of 12
122-00450.45 μmSFCA50 mm115 mLCase of 72
122-0045PK0.45 μmSFCA50 mm115 mLPack of 12
155-00200.2 μmSFCA50 mm150 mLCase of 12
155-00450.45 μmSFCA50 mm150 mLCase of 12
157-00200.2 μmSFCA50 mm250 mLCase of 12
157-00450.45 μmSFCA50 mm250 mLCase of 12
156-40200.2 μmSFCA75 mm500 mLCase of 12
162-00200.2 μmSFCA90 mm500 mLCase of 12
156-40450.45 μmSFCA75 mm500 mLCase of 12
162-00450.45 μmSFCA90 mm500 mLCase of 12
158-00200.2 μmSFCA75 mm500 mLCase of 12
161-00200.2 μmSFCA90 mm1000 mLCase of 12
158-00450.45 μmSFCA50 mm500 mLCase of 12
161-00450.45 μmSFCA90 mm1000 mLCase of 12
150-00200.2 μmNylon50 mm150 mLCase of 12
150-00450.45 μmNylon50 mm150 mLCase of 12
153-00200.2 μmNylon50 mm250 mLCase of 12
153-00450.45 μmNylon50 mm250 mLCase of 12
151-40200.2 μmNylon75 mm500 mLCase of 12
163-00200.2 μmNylon90 mm500 mLCase of 12
151-40450.45 μmNylon75 mm500 mLCase of 12
154-00200.2 μmNylon75 mm1000 mLCase of 12
164-00200.2 μmNylon90 mm1000 mLCase of 12
154-00450.45 μmNylon75 mm1000mLCase of 12
Catalog number 169-0045
Price (USD)
209.35
Online exclusive
279.13
Save 69.78
Case of 12
Pore Size:
0.45 μm
Material (Membrane):
PES
Diameter (Metric) Membrane:
90 mm
Volume (Metric) Receiver:
500 mL
Unit Size:
Case of 12
Price (USD)
209.35
Online exclusive
279.13
Save 69.78
Case of 12

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ Disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers. Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see Full Specifications in the product table for item participation).

  • Available in PES, CN, SFCA and Nylon membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.
  • Available with Fit for Purpose Sterile documentation
  • Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see "Full Specifications" in the product table for item participation)
Specifications
ACT LabelYes
Diameter (Metric) Membrane90 mm
No. per Case12
No. per Pack12
TypeFilter Unit
Volume (Metric) Receiver500 mL
Volume (Metric) Upper500 mL
DisposableSingle-use
Fit for PurposeSterile
Material (Membrane)PES
Pore Size0.45 μm
SterilitySterile
Unit SizeCase of 12

Frequently asked questions (FAQs)

What is the maximum volume that I can run through the Nalgene Rapid-Flow Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes?

The filters included with Nalgene Rapid-Flow Sterile Single Use Vacuum Filter Units are not specifically rated for a maximum volume that may be filtered. In many cases it depends on the specific application of the filter unit and would require testing beyond the volume of the receiver. Signs of the filter clogging, such as a decrease in filtration rate, would likely indicate the need for replacement.

I am using a Nalgene Rapid-Flow Sterile Disposbable Filter Unit and my solution is taking a long time to filter. Do you have any recommendations to improve filtering?

We recommend using glass fiber prefilters for filtration of solutions with high viscosity or especially high particulate load. The glass fiber prefilters are available as an optional accessory and bundled with select Thermo Scientific Nalgene Rapid-Flow filter units. The prefilters enable a higher volume to be filtered before significant flow decay by trapping and preventing the larger particulate matter from entering the final filter membrane. Simply place one prefilter on top of the membrane bonded inside the unit, wet slightly to hold in place, and proceed with filtration as usual.