Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD157 (BST1) Antibodies

          Invitrogen

          CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), APC, eBioscience™

          2 Published Figures
          2 References
          View all (36) CD157 (BST1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), APC, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (2)
          CD157 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD157 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD157 Antibody (17-1579-42) in Flow

          Staining of normal human peripheral blood cells with Mouse IgG1 K Isotype Control APC (Product # 17-4714-81) (left) or Anti-Human CD157 APC (right). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD157 Antibody in Flow Cytometry (Flow)
          CD157 Antibody in Flow Cytometry (Flow)
          CD157 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          17-1579-42

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          5 µL (0.125 µg)/test
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioSY11B5 (SY11B5)

          Conjugate

          APC APC APC
          • Unconjugated
          • Alexa Fluor 488 
          • NovaFluor Blue 660-120S
          • NovaFluor Red 710
          • NovaFluor Yellow 590
          • NovaFluor Yellow 610
          • PE 
          • PE-Cyanine7
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2573166

          Product Specific Information

          Description: The eBioSY11B5 monoclonal antibody recognizes human CD157 (Mo5, BST-1). CD157 is a 42-45 kDa, GPI-anchored protein with structural and functional similarities with CD38. CD157 was initially cloned because of its expression on monocytes and macrophages, and was subsequently discovered to be the same protein named BST-1, discovered for its expression on bone marrow stromal cells and its ability to stimulate the proliferation of a mouse pre-B cell line. CD157 is a pleiotropic ectoenzyme and is thought to act independently as an enzyme and receptor. Similar to CD38, CD157 is involved in the metabolism of NAD+ and this activity may be involved in regulating intracellular Ca2+ levels. As a receptor, upon binding of its putative ligand, CD157 is thought to initiate a signal transduction cascade resulting in the phosphorylation of cytoplasmic proteins including focal adhesion kinase (FAK). The mechanism and functional significance of CD157-initiated signal transduction remain to be fully characterized.

          Applications Reported: This eBioSY11B5 (SY11B5) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBioSY11B5 (SY11B5) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD157, also known as Bone Marrow Stromal Cell Antigen 1 (BST1), is a pleiotropic ectoenzyme and glycosylphosphatidylinositol-anchored molecule that functions both as an enzyme and a receptor. Similar to CD38, CD157 is involved in the metabolism of NAD+, which may play a role in regulating intracellular Ca2+ levels. As an enzyme, CD157 is also referred to as ADP-ribosyl cyclase 2 and DAMP-1, and it facilitates pre-B cell growth. As a receptor, CD157 is thought to initiate a signal transduction cascade upon binding to its putative ligand, leading to the phosphorylation of cytoplasmic proteins, including focal adhesion kinase (FAK). While the mechanism and functional significance of CD157-initiated signal transduction are not fully characterized, its role in cellular signaling is evident. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis, suggesting a link to polyclonal B cell abnormalities observed in the disease. This overexpression in the stromal cell population may contribute to the pathophysiology of rheumatoid arthritis, highlighting the importance of CD157/BST1 in both normal immune function and disease states.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: ADP-ribosyl cyclase 2; bone marrow stromal antigen 1; cADPr hydrolase 2; CD157 antigen; cyclic ADP-ribose hydrolase 2; NAD(+) nucleosidase; NAD+ nucleosidase; unnamed protein product

          View more View less

          Gene Aliases: cADPR2; CD157

          View more View less

          UniProt ID: (Human) Q10588

          View more View less

          Entrez Gene ID: (Human) 683

          View more View less

          Function(s)
          hydrolase activity, acting on glycosyl bonds NAD+ nucleotidase, cyclic ADP-ribose generating
          Process(es)
          regulation of cell-matrix adhesion regulation of cellular extravasation humoral immune response signal transduction positive regulation of cell proliferation regulation of actin cytoskeleton organization regulation of inflammatory response regulation of calcium-mediated signaling regulation of neutrophil chemotaxis regulation of superoxide metabolic process regulation of integrin-mediated signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0