Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SIINFEKL Antibodies

          Invitrogen

          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), APC, eBioscience™

          18 Published Figures
          41 References
          View all (10) SIINFEKL antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), APC, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (18)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody (17-5743-80) in Flow

          Staining of unpulsed (blue histogram) or SIINFEKL-peptide-pulsed (purple histogram) C57BL/6 splenocytes with 0.06 µg of Anti-Mouse OVA (257-264) (SIINFEKL) peptide bound to H-2Kb APC. Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)
          OVA257-264 (SIINFEKL) peptide bound to H-2Kb Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          17-5743-80

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 41 publications 41 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio25-D1.16 (25-D1.16)

          Conjugate

          APC APC APC
          • Unconjugated
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 615
          • Brilliant UV 737
          • Brilliant Violet 711
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1311288

          Product Specific Information

          Description: The 25-D1.16 monoclonal antibody reacts with the ovalbumin-derived peptide SIINFEKL bound to H-2Kb of MHC class I, but not with unbound H-2Kb, or H-2Kb bound with an irrelevant peptide. This antibody has proven to be very useful tracking the quantity and localization of these specific antigen-presenting cells (APC) in vivo.

          Applications Reported: This eBio25-D1.16 (25-D1.16) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBio25-D1.16 (25-D1.16) antibody has been tested by flow cytometric analysis of SIINFEKL peptide pulsed mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Cells can be pulsed with the SIINFEKL peptide according to the following protocol:

          1. With cells in flow staining buffer, add SIINFEKL peptide to a final concentration of 30 uM.

          2. Incubate cells at 37°C for 2 hours.

          3. Wash cells with flow staining buffer.

          4. Proceed with cell surface staining as normal.

          For additional information see the references listed below.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: H-2Kb-SIINFEKL; OVA-Kb; OVA257-264; OVA257-264 H-2Kb

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0