Keratinocyte SFM (1X) Kit - FAQs
Keratinocyte SFM (1X) Kit - FAQs
View additional product information for Keratinocyte SFM (1X) Kit - FAQs (17005042)
5 product FAQs found
Are the Keratinocyte SFM basal medium and supplement kit available separately?
The basal medium is not available for purchase individually, and can only be ordered as part of the Keratinocyte SFM kit (catalog # 17005-042). The supplements are available separately: Bovine Pituitary Extract is catalog # 13028-014 and the EGF is catalog # PHG0311.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which serum-free media can be used to culture mouse keratinocytes?
The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which serum-free media can be used to culture human keratinocytes?
There are a variety of options for serum-free culture of human keratinocytes. Keratinocyte SFM (Cat. No. 17005-042) is a widely used kit that includes the basal medium, bovine pituitary extract (BPE) and Epidermal Growth Factor (EGF). If you would like a serum-free, defined culture system, the options are the Defined Keratinocyte SFM kit (Cat. No. 10744-019) or Epilife medium (Cat. No. M-EPI-500-CA) supplemented with EDGS (Cat. No. S-012-5). If you would like an animal origin free defined system, Epilife medium (Cat. No. M-EPI-500-CA) with S7 supplement (Cat. No. S-017-5) is an excellent choice.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the calcium concentration in Keratinocyte SFM (1X) (Cat. No. 17005042)?
Keratinocyte SFM (1X) (Cat. No. 17005042) includes Keratinocyte-SFM (Cat. No. 10724011) and Keratinocytes Supplements (Cat. No. 37000015), which contains Bovine Pituitary Extract (BPE) EGF, Human Recombinant. The complete media is prepared by adding the supplements to the base media. The Keratinocyte-SFM (Cat. No. 10724011) contains calcium chloride at 12.98 mg/L and D calcium pantothenate at 0.29 mg/L.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
For how long can I store Keratinocyte-SFM at 4 degrees C?
The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.