2 mL Serological Pipettes, paper/plastic peel, individually wrapped, Pack of 125 - FAQs
2 mL Serological Pipettes, paper/plastic peel, individually wrapped, Pack of 125 - FAQs
View additional product information for Nunc™ Serological Pipettes - FAQs (170374N, 170373N, 170372N, 170371N, 170358N, 170369N, 170357N, 170368N, 170356N, 170367N, 170355N, 170366N, 170354N, 170365N, 170376N, 170353N, 170364N, 170375N)
2 product FAQs found
Do Nunc Serological Pipettes have an aspirator plug at the top?
Yes, Nunc Serological Pipettes have an aspirator plug at the top.
What is the difference in between Nunc Serological Pipettes and Cat. No. 170354N and Cat. No. 170354?
The Nunc Serological Pipettes (Cat. No. 170354N) are the New (N) version of the Nunc Serological Pipettes (Cat. No. 170354). They have the same design. Cat. No. 170354N has an additional certification since it is manufactured at a different site than the older version (Cat. No. 170354). Cat. No. 170354N is certified Non-Cytotoxic, RNAse- and DNAse-fre, and the PET filter is Human DNA-Free. The older version, Cat. No. 170354 is certified non-cytotoxic and non-pyrogenic, and the filter is made of cotton.