10 mL Serological Pipettes, paper/plastic peel, individually wrapped, Pack of 100 - FAQs
10 mL Serological Pipettes, paper/plastic peel, individually wrapped, Pack of 100 - FAQs
View additional product information for Nunc™ Serological Pipettes - FAQs (170374N, 170373N, 170372N, 170371N, 170358N, 170369N, 170357N, 170368N, 170356N, 170367N, 170355N, 170366N, 170354N, 170365N, 170376N, 170353N, 170364N, 170375N)
1 product FAQs found
Do Nunc Serological Pipettes have an aspirator plug at the top?
Yes, Nunc Serological Pipettes have an aspirator plug at the top.