D-Alanine, 99+%
D-Alanine, 99+%
D-Alanine, 99+%
Thermo Scientific Chemicals

D-Alanine, 99+%

CAS: 338-69-2 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 172080050
Price (USD)
51.40
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
51.40
Each
Chemical Identifiers
CAS338-69-2
IUPAC Name(2R)-2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-UWTATZPHSA-N
SMILESC[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 % (On dry substance)
Loss on drying=<0.2 % (105°C, 3 hrs)
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only