tert-Butyl carbamate, 98%
tert-Butyl carbamate, 98%
tert-Butyl carbamate, 98%
Thermo Scientific Chemicals

tert-Butyl carbamate, 98%

CAS: 4248-19-5 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 173450050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS4248-19-5
IUPAC Nametert-butyl carbamate
Molecular FormulaC5H11NO2
InChI KeyLFKDJXLFVYVEFG-UHFFFAOYSA-N
SMILESCC(C)(C)OC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals or needles
Melting point106°C to 110°C
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only