alpha-D-Methylglucoside, 99%
alpha-D-Methylglucoside, 99%
alpha-D-Methylglucoside, 99%
Thermo Scientific Chemicals

alpha-D-Methylglucoside, 99%

CAS: 97-30-3 | C7H14O6 | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 173840025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS97-30-3
IUPAC Name2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular FormulaC7H14O6
InChI KeyHOVAGTYPODGVJG-UHFFFAOYNA-N
SMILESCOC1OC(CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<0.1 % (60 to 70°C)
Specific optical rotation+156° to +161° (20°C, 589 nm) (c=10, water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only