Formamide
Thermo Scientific™

Formamide

Thermo Scientific Pierce Formamide is a high-purity formulation of formamide denaturant that is suitable for the preparation of hybridization cocktailsRead more
Have Questions?
Catalog NumberQuantity
17899100 mL
Catalog number 17899
Price (USD)
134.00
Each
Quantity:
100 mL
Price (USD)
134.00
Each
Thermo Scientific Pierce Formamide is a high-purity formulation of formamide denaturant that is suitable for the preparation of hybridization cocktails and various denaturing solutions used with nucleic acids.

Features of Formamide:

• Ultra pure formulation possesses a high degree of purity and is free of contaminating RNase and DNase
• Suitable for the preparation of hybridization cocktails and various denaturing solutions used with nucleic acids
• Will not cause degradation of expensive or hard to isolate RNA and DNA samples
• Pre-filtered through a mixed-ion exchange resin to eliminate products of decomposition and residual metal cations
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialFormamide
Product TypeFormamide
Quantity100 mL
FormLiquid
Unit SizeEach
Contents & Storage
Store at 2–8°C.