Proteinase K Powder (≥30 U/mg) - FAQs
Proteinase K Powder (≥30 U/mg) - FAQs
View additional product information for Proteinase K Powder (≥30 U/mg) - FAQs (17916)
1 product FAQs found
How do you recommend storing proteinase K?
We recommend storing it at 4°C.