Thermo Scientific Pierce Tris is a standard reagent for preparation of many molecular biology buffers.Features of Thermo Scientific Pierce Tris•자세히 알아보기
Have Questions?
카탈로그 번호
수량
17926
1 kg
카탈로그 번호 17926
제품 가격(KRW)
204,000
온라인 행사
Ends: 30-Jun-2026
240,000
할인액 36,000 (15%)
Each
수량:
1 kg
제품 가격(KRW)
204,000
온라인 행사
Ends: 30-Jun-2026
240,000
할인액 36,000 (15%)
Each
Thermo Scientific Pierce Tris is a standard reagent for preparation of many molecular biology buffers.
Features of Thermo Scientific Pierce Tris
• ≥99.8% pure • White crystalline powder • Optimal buffering between pH 7 and 9
For Research Use Only. Not for use in diagnostic procedures.
사양
제품 유형Tris
순도≥99.8%
수량1 kg
형태Powder
Unit SizeEach
구성 및 보관
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.