eBioscience™ Avidin HRP, 1 mL - FAQs
eBioscience™ Avidin HRP, 1 mL - FAQs
View additional product information for eBioscience™ Avidin HRP - FAQs (18-4100-94, 18-4100-51)
1 product FAQs found
What other reagents will I need to run an ELISA using the antibody pairs?
When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.