Ribonuclease H, 30 U - FAQs
Ribonuclease H, 30 U - FAQs
View additional product information for Ribonuclease H - FAQs (18021071, 18021014)
2 product FAQs found
Do you offer RNAse H from the SuperScript III First Strand Synthesis System as a stand-alone item?
Yes, we do offer E. coli RNAse H as a stand-alone item: Cat. Nos. 18021014 and 18021071.
Which components of the SuperScript III First Strand Synthesis System for RT-PCR are available for purchase separately?
The following components are available as stand-alone items:
- Superscript III Reverse Transcriptase (Cat. Nos. 18080093, 18080044, 18080085)
- Oligo (dT)20 Primer (Cat. No. 18418020)
- Random hexamers (Cat. No. 48190011)
- 10 mM dNTP Mix (Cat. Nos. 18427013, 18427088)
- RNAseOUT Recombinant Ribonuclease Inhibitor (Cat. No. 10777019)
- E. coli RNAse H (Cat. Nos. 18021014, 18021071)