Tryptose Phosphate Broth - FAQs
Tryptose Phosphate Broth - FAQs
View additional product information for Tryptose Phosphate Broth - FAQs (18050039)
1 product FAQs found
What is the concentration of tryptose phosphate solution in Tryptose Phosphate Broth (Cat. No. 18050039)?
The Tryptose Phosphate Broth contains 29.5 g/L tryptose phosphate.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.