SuperScript™ Choice System for cDNA Synthesis - FAQs
SuperScript™ Choice System for cDNA Synthesis - FAQs
View additional product information for SuperScript™ Choice System for cDNA Synthesis - FAQs (18090019)
1 product FAQs found
I'd like to generate double-stranded DNA (dsDNA) from mRNA. Do you have a kit for this?
To generate double-stranded cDNA, we recommend the Maxima H Minus Double-Stranded cDNA Synthesis Kit (Cat. No. K2561) or Second Strand cDNA Synthesis Kit (Cat. No. A48570).