Can a single-stranded DNA be generated using M13 helper phage?
Yes. Use a strain that expresses endonuclease A (e.g., DH12S, DH11S). The orientation of the single-stranded DNA will be the opposite of that generated by Gene II and Exo III.
Search
View additional product information for ElectroMAX™ DH12S Cells - FAQs (18312017)
2 product FAQs found
Yes. Use a strain that expresses endonuclease A (e.g., DH12S, DH11S). The orientation of the single-stranded DNA will be the opposite of that generated by Gene II and Exo III.