SuperScript V Reverse Transcriptase
Invitrogen™

SuperScript V Reverse Transcriptase

Invitrogen™ SuperScript™ V Reverse Transcriptase (RT) is a next-generation MMLV reverse transcriptase designed for fast, reliable first-strand cDNA synthesis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
190900102000 Units
1909005010,000 Units
1909020040,000 Units
Catalog number 19090010
Price (USD)
97.00
Each
Quantity:
2000 Units
Price (USD)
97.00
Each

Invitrogen™ SuperScript™ V Reverse Transcriptase (RT) is a next-generation MMLV reverse transcriptase designed for fast, reliable first-strand cDNA synthesis. Building on the advanced performance of the SuperScript™ IV reverse transcriptase, SuperScript V RT combines high sensitivity, thermostability, and processivity with improved resistance to oxidation and greater workflow flexibility.

SuperScript V RT supports reliable cDNA synthesis from low-input RNA as well as highly structured, GC-rich, degraded, and inhibitor-containing samples.

Features of SuperScript V Reverse Transcriptase include:

  • Broad RNA input range—supports cDNA synthesis from 100 fg to 5 μg of RNA
  • Robust performance with challenging RNA—reaction temperatures up to 65°C help transcribe highly structured and GC-rich RNA
  • Fast cDNA synthesis—complete first-strand cDNA synthesis in as little as 1–10 minutes
  • Improved resistance to oxidation— enhanced reagent stability supports ambient shipping and room-temperature storage for up to 6 months
  • Flexible workflow—room-temperature reaction setup and stability of assembled reactions for up to 24 hours

SuperScript V RT is well suited for RT-PCR, RT-qPCR, gene expression analysis, and other applications requiring high-quality first-strand cDNA synthesis. Its enhanced reagent and assembled-reaction stability provides additional flexibility for routine, high-throughput, and automated workflows.

A DTT-free, animal origin–free formulation combined with reduced cold chain requirements further simplifies reagent handling and supports more sustainable workflows.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatStand-alone Enzyme
Inhibitor Tolerance (Simple)High
No. of Reactions10 Reactions
Optimal Reaction Temperature50°C
Product TypeReverse Transcription Reagent
Quantity2000 Units
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseSuperScript V
Ribonuclease H ActivityReduced
Sensitivity0.1 pg total RNA
Shipping ConditionDry Ice, Wet Ice, Ambient
Size (Final Product)Up to 20 kb
Starting MaterialRNA
TechniqueReverse Transcription
ThermostabilityUp to 65°C
Concentration200 U/μL
Detection MethodqPCR, NGS, PCR
For Use With (Application)cDNA Synthesis, RT-PCR, Real Time PCR (qPCR), Sequencing, Next-Generation Sequencing (NGS)
FormLiquid
GC-Rich PCR PerformanceHigh
Label or DyeNot Applicable
PCR Method2-step RT-qPCR, RT-PCR
Reaction Speed10 min.
Unit SizeEach
Contents & Storage
• SuperScript V Reverse Transcriptase (10 μL)
• 5X SuperScript V Reaction Buffer (500 μL)

Store all contents at:
• -20°C for long-term storage until expiry date
• +5°C for up to 12 months
• Room temperature for up to 6 months

Frequently asked questions (FAQs)

What are the main differences between Invitrogen SuperScript V Reverse Transcriptase and Invitrogen SuperScript V VILO Master Mix?

Invitrogen SuperScript V Reverse Transcriptase is supplied as a stand-alone enzyme with reaction buffer and supports flexible reaction optimization. Invitrogen SuperScript V VILO Master Mix combines all required reaction components into a single formulation to simplify workflow setup and reduce pipetting steps.

Can Invitrogen SuperScript V Reverse Transcriptase products support automated workflows?

Yes. Invitrogen SuperScript V Reverse Transcriptase products include features such as room-temperature reaction setup, benchtop stability of assembled reactions, and simplified workflow options that can support automated and high-throughput workflows.

Can Invitrogen SuperScript V Reverse Transcriptase products support low-input RNA workflows?

Yes. Invitrogen SuperScript V Reverse Transcriptase products are designed to support reverse transcription from low-input RNA samples, including inputs as low as 0.1 pg of total RNA.

Which Invitrogen SuperScript V Reverse Transcriptase product should I choose for RT-qPCR workflows?

For two-step RT-qPCR workflows, Invitrogen SuperScript V VILO Master Mix reagent (Cat. No. 11800001) provides an all-in-one formulation that simplifies reaction setup. Users who prefer custom primer selection may choose Invitrogen SuperScript V VILO Master Mix, Primer-Free (Cat. No. 11900001) or Invitrogen SuperScript V Reverse Transcriptase (Cat. No. 19090050).

Can reaction volumes be scaled when using Invitrogen SuperScript V Reverse Transcriptase products?

Yes. Reaction volumes may be scaled up to 100 µL when using Invitrogen SuperScript V Reverse Transcriptase products provided that reagent ratios are maintained and workflow performance is verified.