Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, GMP, J.T.Baker™
Avantor™

Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
1919-08200 lb.Fiber Drum
1919-09100 lb.Fiber Drum
1919-01500 gPoly Bottle, Tamper-evident Cap
1919-25200 lb.Polyethylene Drum, Open Head
1919-052.5 kgPoly Bottle, Tamper-evident Cap
1919-0712 kgPolyethylene Pail, Screw Top
Catalog number 1919-08
Price (USD)
-
Quantity:
200 lb.
Packaging:
Fiber Drum
This product is GMP Manufactured and meets USP specifications.
This product is GMP Manufactured and meets USP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorWhite
Melting Point146°C
PackagingFiber Drum
GradeUSP
Molecular Weight (g/mol)180.16
Percent Purity97.5 to 102%
Physical FormSolid
Quantity200 lb.
Unit SizeEach