Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder
Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder
Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder
Thermo Scientific Chemicals

Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder

CAS: 7440-18-8 | Ru | 101.07 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 198100010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
さらに表示
Total trace metal impurities=<0.1 %
Appearance (Color)Grey
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only