ViewRNA™ Tissue Assay Core Kit - FAQs
ViewRNA™ Tissue Assay Core Kit - FAQs
View additional product information for ViewRNA™ Tissue Assay Core Kit - FAQs (19931)
4 product FAQs found
Are there any changes in the reagents and buffers used in the ViewRNA Tissue Assay Core Kit relative to the discontinued ViewRNA ISH Tissue Assay kits?
No. The reagents and buffers used in the ViewRNA Tissue Assay Core Kit are the same as the ones used in the discontinued ViewRNA ISH Tissue Assay kits.
Can I purchase the individual components of the ViewRNA Tissue Assay Core Kit and ViewRNA Tissue Assay Blue Module as standalone products?
Yes. Please refer to the catalog numbers listed on pages 4 and 5 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018633_viewRNA_ISH_UG.pdf).
How is ViewRNA Tissue Assay Core Kit different from the discontinued ViewRNA ISH Tissue Assay kits?
The discontinued ViewRNA ISH Tissue Assay kits (Cat Nos. QVT0012 and QVT0013) were provided as complete 2-plex kits. Currently, we offer the ViewRNA Colorimetric Tissue Assays in three formats: Core Kit and Individual Modules for Fast Red, Fast Blue, and DAB signal development, 1-plex assay kits, and 2-plex assay Kits. Please find associated product links below:
ViewRNA Colorimetric Tissue Core Kit and Individual Modules
ViewRNA Colorimetric Tissue 1-Plex Kits
ViewRNA Colorimetric Tissue 2-Plex Kits
Label Probe Diluent is provided in the ViewRNA Tissue Assay Core Kit but not in the ViewRNA Tissue Assay Blue Module. Can I purchase this separately?
Yes. It is offered separately as Label Probe Diluent QF, Cat no. QVT0513.