Partisol™ 2000-D Dichotomous Ambient Particulate Sampler
Thermo Scientific™

Partisol™ 2000-D Dichotomous Ambient Particulate Sampler

This product has been discontinued however we continue to provide services and support for this product.
Have Questions?
카탈로그 번호설명
2000DPartisol 2000-D Dichotomous Air Sampler
카탈로그 번호 2000D
제품 가격(KRW)
-
설명:
Partisol 2000-D Dichotomous Air Sampler

This product has been discontinued however we continue to provide services and support for this product.

The Partisol 2000-D simultaneously measures PM-2.5 and PM-Coarse mass concentrations on two separate 47mm cassettes housed on a single action filter change mechanism.

Key Features:

  • US EPA PM-2.5 and PM-Coarse FEM designation
  • Utilizes Filter Cassette Carrier for both filters to eliminate contamination
  • Quick and convenient flow and audit procedures with built-in support for the Streamline Pro System
  • Onboard microprocessor provides large internal data storage buffer

Standard System Configuration:

  • Single unit consisting of manual sampler, stand and sampling inlet
  • PM-10 inlet combined with Virtual Impactor allows for split fine and coarse flow
  • ActiVol automatic flow control utilizing two independent flow controllers
  • RPCOMM software for local and remote interaction
  • Standard 47mm filter cassettes with convenient filter change mechanism

Includes:

  • Consumables for average first year's operation
  • Sampler, stand, pump, and accessories

Compatible with:

  • Streamline Pro
  • Streamline FTS

Recommended for:

  • Air quality monitoring networks, including background sites
  • Special studies and super-sites
  • Routine input for air quality index
  • Remediation projects (Superfund, hazardous waste)
  • In and around industrial and material handling facilities
사양
암페어0.2 A (at 120 VAC); 1.1 A (at 240 VAC)
인증/적합성USEPA PM-2.5 Equivalent sampler: EQPS-0509-177
데이터 포인트12 days of Interval data (stored every 5 minutes)
깊이(영국식 단위)13 in.
깊이(미터법)33 cm
설명Partisol 2000-D Dichotomous Air Sampler
필터 유형47 mm diameter. Partisol-FRM style filter cassette
높이(영국식 단위)24 in.
높이(미터법)61 cm
출력Display screen, analog output (0-5VDC) of status and flow, RS232.
샘플 흐름 제어 및 보고Air flow through the sampler is provided by a proven low-maintenance vacuum pump. Activol flow control system uses mass flow sensors and the measured ambient temperature and pressure to maintain constant volumetric flow rates. System flow rate is 16.7 L/min. (1 m3/hr).
온도(영국식 단위) 작동-22°F to +122°F
중량(영국식 단위)70 lb.
중량(미터법)32 kg
폭(미터법)41 cm
작동 온도(미터법) 작동-30°C to +50°C
Unit SizeEach