Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure
Thermo Scientific™

Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure

Store, ship and package liquids with these highly reliable, lab-quality bottles, specifically designed for long-term demanding lab use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2002-9016PK500 mL38-430 mmPack of 12
2002-91254 mL13 mmCase of 72
2002-9125PK
also known as 2002-9125
4 mL13 mmPack of 12
2002-90258 mL20 mmCase of 72
2002-9025PK
also known as 2002-9025
8 mL20 mmPack of 12
2002-905015 mL20 mmCase of 72
2002-9050PK15 mL20 mmPack of 12
2002-000130 mL20 mmCase of 72
2002-0001PK30 mL20 mmPack of 12
2002-000260 mL20 mmCase of 72
2002-0002PK
also known as 2002-0002
60 mL20 mmPack of 12
2002-0004125 mL24 mmCase of 72
2002-0004PK
also known as 2002-0004
125 mL24 mmPack of 12
2002-0006175 mL24 mmCase of 72
2002-0006PK
also known as 2002-0006
175 mL24 mmPack of 12
2002-0008250 mL24 mmCase of 72
2002-0008PK
also known as 2002-0008
250 mL24 mmPack of 12
2002-9016500 mL38-430 mmCase of 48
2002-0016500 mL28 mmCase of 48
2002-0016PK
also known as 2002-0016
500 mL28 mmPack of 12
2002-00321 L38-430 mmCase of 24
2002-0032PK1 L38-430 mmPack of 6
Catalog number 2002-9016PK
Price (USD)
103.23
Online exclusive
114.70
Save 11.47
Pack of 12
Capacity (Metric):
500 mL
Closure Size:
38-430 mm
Unit Size:
Pack of 12
Price (USD)
103.23
Online exclusive
114.70
Save 11.47
Pack of 12

Store, ship, and package liquids using highly reliable and durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure. These semi-rigid, translucent, all-purpose bottles are specifically designed for long-term, demanding lab use and offer excellent chemical resistance. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

  • Versatile and indispensable in the lab
  • Uniform walls provide durability and resistance to splitting or punctures
  • Can be used in a freezer up to -100°C (-149°F)
  • Linerless polypropylene closures ensure leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity16 oz., 500 mL
Capacity (English)16 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorNatural
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38-430 mm
Closure TypeScrew Cap
DescriptionLab Quality Narrow-Mouth Bottle
Diameter (Metric)73 mm
Diameter (Metric) Inner Neck27 mm
Height (English)6.69 in.
Height (Metric)170 mm
Neck Finish38 mm
No. per Case48
No. per Pack12
Product TypeLaboratory Bottle
ShapeRound
Through the Closure DispensingNo
Capacity (Metric)500 mL
Closure IncludedYes
ColorTranslucent
MaterialHDPE
MouthNarrow
SterilityNon-sterile
Unit SizePack of 12

Frequently asked questions (FAQs)

I need a polypropylene product but all I can find is polypropylene copolymer. What‘s the difference?

At the molecular level, polypropylene copolymer consists of a pattern of co-polymerized ethylene and propylene monomer “units” and so in some ways could be considered a “mixture” of polyethylene and polypropylene. It was created in order to combine some of the ideal properties of each; i.e., the high-temperature warping resistance of polypropylene and the low-temperature brittleness resistance of polyethylene. The polymer industry's standard nomenclature for this material is polypropylene copolymer. In common parlance, however, “polypropylene” can often refer to copolymer (PPCO) as well as “pure” polypropylene (homopolymer, PP or PPHO) consisting entirely of propylene monomers. Many of the product molds we use for PPCO items were set up to stamp a “PP” material specification on the base, which can be misleading. Our specification sheets and compliance certificates, however, always accurately identify the material in question.