Nalgene™ Rectangular HDPE Bottles with Closure
Nalgene™ Rectangular HDPE Bottles with Closure
Nalgene™ Rectangular HDPE Bottles with Closure
Thermo Scientific™

Nalgene™ Rectangular HDPE Bottles with Closure

Transport, sample or store liquids and dry materials with these space-efficient bottles featuring excellent chemical resistance to most acids, bases and alcohols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2007-00321 L53 mmCase of 24
2007-0004125 mL28 mmCase of 72
2007-0004PK
also known as 2007-0004
125 mL28 mmPack of 12
2007-0008250 mL38 mmCase of 72
2007-0008PK
also known as 2007-0008
250 mL38 mmPack of 12
2007-0016500 mL48 mmCase of 48
2007-0016PK
also known as 2007-0016
500 mL48 mmPack of 12
2007-0032PK
also known as 2007-0032
1 L53 mmPack of 6
2007-00642 L63 mmCase of 12
2007-0064PK
also known as 2007-0064
2 L63 mmPack of 4
Catalog number 2007-0032
Price (USD)
309.68
Online exclusive
397.70
Save 88.02
Case of 24
Capacity (Metric):
1 L
Closure Size:
53 mm
Unit Size:
Case of 24
Price (USD)
309.68
Online exclusive
397.70
Save 88.02
Case of 24

Transport, sample or store liquids and dry materials with these space-efficient bottles that feature excellent chemical resistance to most acids, bases and alcohols. Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure take up less shelf space than traditional round bottles. HDPE bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Rectangular shape makes it easy to fit in freezer for storage up to -100°C (-148°F)
  • Linerless polypropylene screw closures provide leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability

Includes:

Polypropylene screw caps

Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
ACT LabelYes
Capacity32 oz., 1000 mL
Capacity (English)32 oz.
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure ColorNatural, translucent
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size53 mm
Closure TypeScrew Cap
DescriptionWide-Mouth Rectangular Bottle
Diameter (English) Inner Neck1.7 in.
Diameter (Metric)124.5 mm
Diameter (Metric) Inner Neck44 mm
Height (English)7 in.
Height (Metric)180 mm
Length (Metric)125 mm
No. per Case24
No. per Pack6
Product TypeWide Mouth Bottle
ShapeRectangular
Through the Closure DispensingNo
Width (Metric)71 mm
Capacity (Metric)1 L
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialHDPE
MouthWide
SterilityNon-sterile
Unit SizeCase of 24

Frequently asked questions (FAQs)

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.