LightShift™ Chemiluminescent EMSA Kit - FAQs
LightShift™ Chemiluminescent EMSA Kit - FAQs
View additional product information for LightShift™ Chemiluminescent EMSA Kit - FAQs (20148)
43 product FAQs found
If the LightShift Chemiluminescent EMSA kit has been improperly stored (i.e., at room temperature, -20°C or +4°C), will it still work correctly?
The LightShift Chemiluminescent EMSA Kit is composed of two sets of components that require different storage temperatures. One component set consists of the chemiluminescent substrates and various buffers that are stored at 4°C. The other component set consists of the control DNAs and various optimization reagents that are stored at -20°C. The EBNA extract must be maintained at -20°C or it will lose activity (proteins will degrade). Short-term storage (overnight) of the other kit components at temperatures ranging from room temperature to -20°C will not adversely affect kit performance.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am using the LightShift Chemiluminescent EMSA kit. Can I probe for the proteins by performing a Western blot?
This has not been tested but may be possible. A better alternative is to perform a DNA binding protein pull-down assay using a probe. The following journal article is a good example of how the LightShift Chemiluminescent EMSA Kit and pull-down assays were used to detect a transcription factor bound to a DNA probe: Ragione, A.L., et al. (2003), J. Biol. Chem. 278(26):23360-8.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am using the LightShift Chemiluminescent EMSA kit. How much protein do I need for each reaction?
The amount of protein extract needed for a binding reaction depends on how much active DNA binding protein is in the sample. The LightShift Kit is sensitive and will easily detect 5 fmol of active protein bound to 5 fmol of biotinylated probe. If the protein being studied is abundant, 0.25 µg of a cell lysate may be sufficient for each binding reaction. However, if the protein of interest is rare, 10 µg or more of cell lysate may be needed. Using a large excess of protein extract may lead to high background signal and non-specific bands.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is a "supershift"?
A supershift assay is a method for positively identifying a protein:DNA interaction on an EMSA. An antibody (typically 1 µg) is added to the binding reaction. During electrophoresis, the antibody:protein:DNA complex migrates slowly, causing a “supershift” compared to the “shift” caused by a protein:DNA complex. Not all antibodies will cause a supershift. Some antibodies do not bind to proteins once they are bound to DNA. Some antibodies can prevent protein:DNA interactions but can still be used to confirm the identity of a protein that causes a shift in the absence of the antibody.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can the LightShift Chemiluminescent EMSA Kit be used to perform supershifts?
Yes, the LightShift Chemiluminescent EMSA Kit can be used to detect supershifts. However, not all antibodies will work for supershift assays. Some antibodies will prevent protein:DNA interactions. In addition, the order in which the components of the binding reaction are assembled may affect the results of a supershift assay. Generally, 1 µg antibody is added as the last component in the binding reaction. For examples of how the LightShift Chemiluminescent EMSA Kit was used to detect supershifts, see the following:
Adimoolam, S. and Ford, J.M. (2002). PNAS. 99(20):12985-90
Magid, R., et al. (2003). J. Biol. Chem. 278(35):32994-9
Ragione, F.D., et al. (2003). J. Biol. Chem. 278(26):23360-8
Rinaldi, A.L., et al. (2002). Cancer Research. 62(19):5451-6
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the purpose of adding unlabeled DNA to the binding reaction as a control in the LightShift Chemiluminescent EMSA kit?
As a control, unlabeled DNA of the same sequence as the biotin-labeled DNA is added in excess to compete with the biotin-labeled DNA. This results in a decrease or elimination of the “shift”, verifying that the protein-DNA interaction was sequence specific.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am using the LightShift Chemiluminescent EMSA kit. What are the optimal reaction conditions for my protein?
The binding conditions for each experiment must be optimized empirically for the proteins tested. Journal articles are an excellent source for information on how to set up EMSA binding reactions. If an article describes an EMSA for your protein of interest but used 32P labeled probes, repeat the binding conditions with biotinylated probe in place of the 32P labeled probe. Although reaction components are provided with the LightShift Chemiluminescent EMSA Kit, these reagents may not be suitable for individual applications and may be adjusted as needed.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am using the LightShift Chemiluminescent EMSA kit. Why is my shift not blocked by "cold" (unlabeled) probe?
The shift may be caused by non-specific binding or a DNA binding protein different than the one being tested.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am using the LightShift Chemiluminescent EMSA kit. How do I make the non-specific bands go away?
Non-specific bands may be reduced or eliminated by optimizing the binding reactions. Strong non-specific bands can often be mistaken for a positive result but will not be blocked by the addition of cold competitor probe. The following are examples of changes that MAY help reduce non-specific bands:
• Reduce the amount of protein extract in the binding reaction.
• Increase the amount of non-specific competitor DNA [i.e., poly(dI-dC)·poly(dI-dC)].
• Use a different non-specific competitor DNA [i.e., sonicated salmon sperm DNA or poly(dA-dT)·poly(dA-dT)].
• Preincubate the protein extract with non-specific competitor DNA before adding the biotinylated probe.
• Shorten or redesign the probe used in the experiment.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
With the LightShift Chemiluminescent EMSA kit, what if I see multiple shifts when I analyze the EMSA?
Multiple shifts in an EMSA are most likely caused by the DNA binding to multiple forms of the protein such as a monomer vs. a dimer, or perhaps from multiple factors binding to the same site or to multiple binding sites within the target DNA (especially common with longer duplexes).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
My gel-shift experiment worked when I used P32 but not now with the LightShift Chemiluminescent EMSA Kit. What is wrong?
The LightShift Chemiluminescent EMSA Kit is used to detect a biotinylated probe in an EMSA. The binding conditions for each experiment must be optimized for the proteins tested. If an experiment was working when 32P labeled probes were used, then use the same binding conditions from those previous experiments and only substitute the biotinylated probe in place of the 32P labeled probe. While commonly used reaction components are provided with the LightShift Chemiluminescent EMSA Kit, these reagents may not be applicable in all situations.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What if I don't see a shift with the LightShift Chemiluminescent EMSA assay?
The reaction may need to be optimized. System optimization can be achieved by adding various components supplied with the kit such as KCl, glycerol, MgCl2 and/or detergents and determining their effects on the shift. Comparing the methods used in journal articles that successfully performed EMSA experiments with the proteins you are testing may help with optimization.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How much of a "shift" should I expect to see with the LightShift Chemiluminescent EMSA assay?
The magnitude of mobility shift will vary in different systems and depends upon the abundance and activity of the target protein. Using the control provided with the kit, approximately 50% of the biotin-labeled DNA should be shifted.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What type of membrane should I use to transfer my completed gel-shift after the LightShift Chemiluminescent EMSA Assay?
Positively charged nylon membrane must be used (e.g., Biodyne B Membrane, Cat. No. 77016).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am using the LightShift Chemiluminescent EMSA kit. How should I transfer my completed gel-shift to a membrane?
Transfer to positively charged nylon membrane can be performed using an electrophoretic transfer apparatus.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Are there any considerations I should be aware of when selecting the type of gels to use after the LightShift Chemiluminescent EMSA Assay?
When possible, use a gel system that was used successfully by other researchers to detect your protein of interest. Commercially available DNA retardation gels often work well for EMSA applications. Both the gel type and running buffer composition can influence how well an EMSA works. The following journal article provides an overview of some considerations often overlooked by researchers performing gel-shift assays: Roder, K. and Schwiezer, M. (2001), Biotechnol. Appl. Biochem. 33:209-214.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I use agarose gels or must I use acrylamide gels for the Chemiluminescent EMSA Assay?
EMSAs may be performed with either polyacrylamide or agarose gels depending upon the resolution requirements of the study system. Traditionally, 4-6% non-denaturing polyacrylamide gels are used. If agarose is used, a capillary transfer may be best. Either capillary or electrical transfers can be performed with polyacrylamide gels.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Is there a limit to the length of target DNA I can use with the LightShift Chemiluminescent EMSA kit?
Typically, the target duplex in EMSA is 20-35 bp long. Our scientists have successfully used DNA sequences as long as 60 bp. The actual binding sequence for protein:DNA interactions is frequently 10-15 bp. Longer probes can be used if the binding sequence is unknown or if multiple regulatory regions are being studied.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How should I prepare my biotinylated DNA for the LightShift Chemiluminescent EMSA assay?
DNA labeled on the 5'- or 3'-end can be ordered directly from your oligo supplier or prepared using a biotin end-labeling kit such as the Biotin 3' End-Labeling Kit (Cat. No. 89818). Internal biotin labels may inhibit binding of the DNA binding protein and are not recommended.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can the LightShift Chemiluminescent EMSA kit components be purchased separately?
Yes. The Detection Module (Cat. No. 89880) and Optimization and Control Kit (Cat. No. 20148X) are available separately.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How long does it take to complete the LightShift Chemiluminescent EMSA assay?
The LightShift Chemiluminescent EMSA assay protocol, from making the DNA probe to visualization of the results, can be completed in approximately 5 hours.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How does the sensitivity of the LightShift Chemiluminescent EMSA Kit compare to systems using DIG or P32?
The LightShift Chemiluminescent EMSA Kit is as sensitive as 32P when optimized. In addition, the experiment can be completed in about 5 hours without the need for an overnight exposure often required when using 32P. As with every system, some optimization may be required. The LightShift Chemiluminescent EMSA Kit has superior detection sensitivity compared to DIG detection systems.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How does the LightShift Chemiluminescent EMSA assay work?
Biotin end-labeled DNA containing a putative or known binding site is incubated with a nuclear extract or purified factor. This reaction is then subjected to gel electrophoresis on a native polyacrylamide gel and transferred to a nylon membrane. Because DNA-protein complexes migrate slower than DNA alone in a native gel, a “shift” in the migration of the labeled DNA occurs. The biotin end-labeled DNA is detected using a streptavidin-horseradish peroxidase conjugate and a chemiluminescent substrate developed for the LightShift Kit. The signal is then detected with X-ray film or a high quality CCD camera.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What are the applications for the LightShift Chemiluminescent EMSA Kit?
The LightShift Chemiluminescent EMSA Kit uses proprietary chemiluminescent detection technology to perform nonradioactive EMSA or supershift assays.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is an electrophoretic mobility shift assay (EMSA)?
EMSAs (also called gel shifts, band shifts, gel retardation assays, or mobility assays) have been used extensively for studying protein-DNA interactions. Because protein-DNA complexes migrate more slowly through a native polyacrylamide or agarose gel than DNA alone, individual protein-DNA complexes can be visualized as discrete bands within the gel using chemiluminescence or radioisotopic detection.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Is the 5X Loading Buffer within the LightShift Chemiluminescent EMSA Kit Cat. No. 20148) stable at room temperature?
The 5X Loading Buffer within the LightShift Chemiluminescent EMSA Kit Cat. No. 20148), ideally, should be stored at -20°C. However, this loading buffer is not very temperature sensitive and even if stored for several days at RT, it should not affect its performance.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.
How do I identify if my protein has bound to my EMSA probe?
You can identify the presence of a specific protein in a shifted band using a supershift assay. An antibody that is specific for the protein of interest is added to the binding reaction. Binding of the antibody to the protein causing the gel shift will often result in decrease in the mobility of the shifted complex, although other effects are possible (such as the disappearance of the shifted band). The order of addition of the components for a supershift is often critical; generally the antibody should be added last. We suggest this order:
- Water, buffer, any additives
- Poly dI-dC
- Nuclear extract (allows for non-specific proteins to bind to the poly dI-dC competitor)
- Biotinylated-DNA duplex
- Antibody (we use about 1 µg)
Incubate for 20 min.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I do not see a shifted band on my EMSA gel. What should I do?
We recommend running run a cold probe competition lane (cold probe + label probe ++ nuclear extract) to see which band becomes fainter when compared to the label probe (label probe + nuclear extract) lane. With the cold probe competition lane, you can better distinguish which is the shifted band of interest.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Which substrate do you recommend using with the LightShift Chemiluminiscent EMSA Kit and LightShift Chemiluminiscent RNA EMSA Kit?
We recommend using the Chemiluminescent Nucleic Acid Detection Module Kit, Cat. No. 89880 as it has been optimized with the control probes in these kits. Results with other HRP-compatible substrates may vary depending on the quality or characteristic of the chemiluminescent substrate used.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I use alkaline transfer with EMSA?
Alkaline transfer contains NaOH and is typically used for northern and Southern blots. We do not recommend alkaline transfer for EMSA. We recommend the use of 0.5X TBE buffer for transfer of EMSA binding reactions. TBE transfer is near-neutral pH condition.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I use PVDF or nitrocellulose membranes for transfer of EMSA binding reactions?
We recommend using positively-charged nylon membranes for transfer of EMSA reactions. Nitrocellulose membranes and PVDF membranes are not ideal as they strongly bind to protein but only weakly bind to DNA.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I use pre-cast gels for running EMSA binding reactions?
We recommend using a native 4-6% polyacrylamide gel in 0.5X TBE buffer. The appropriate polyacrylamide percent depends on the size of the target DNA/RNA and the binding protein. Our precast 6% DNA Retardation gels may be used.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What are the controls provided with your EMSA kits?
The control extract provided is E. coli expressing recombinant EBNA, and the control probe is the Biotin-EBNA Control DNA. These controls should be run together. You can use the control extract, control probe, and cold control probe to affirm the binding reaction, running of the gel, transfer, and detection steps of the protocol.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I make my EMSA probe using PCR?
You can generate the EMSA probe using PCR with 5'-biotin modification. Avoid using probes that are internally labeled with biotin as this may interfere with the binding of target proteins.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How should I anneal oligos for an EMSA assay?
Many different methods can be used to anneal complementary oligo pairs. Please refer to our Tech Tip - Anneal complementary pairs of oligonucleotides: https://tools.thermofisher.com/content/sfs/brochures/TR0045-Anneal-oligos.pdf
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What should I use for diluting the EMSA probes?
The EMSA probes should be diluted in dH2O and stored at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What are your recommendations for designing and labeling of EMSA probes?
Here are our recommendations:
- Typically, end-labeled, double-stranded DNA probes of 20-35 bp length are used, but other sizes are possible (note that the EBNA control probe in the kit is 60 bp long).
- Probes can be ordered with a 5'-biotin modification. They can be also enzymatically labeled on the 3' end using the Pierce Biotin 3' End DNA Labeling Kit (Cat. No. 89818).
- Single-stranded labeled probes should be annealed.
- Probes shorter than 20 bp may not bind the target protein well; longer probes can work but may require additional optimization or adjustments to gel running conditions.
- It is critical to have positive and negative controls in the EMSA assay.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I transfer EMSA gels using the Power Blotter System/Power Blotter XL System?
Yes. You may use the same method that was used with the G2 Fast Blotter: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-biology-learning-center/protein-biology-resource-library/protein-biology-application-notes/transfer-emsa-gels-using-g2-fast-blotter.html
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
With the Lightshift Chemiluminescent EMSA assay, at times, I do not get any signal or the signal is low. How can I prevent this?
Here are some suggestions to improve the results:
  • Make sure that the target DNA used is end-labeled with biotin.
  • Increase target DNA concentration.
  • Check integrity of target DNA to make sure it is not degraded.
  • Check the transfer protocol to make sure that the transfer is efficient.
  • Use the right type of membrane such as Biodyne B Nylon Membrane (Cat. No. 88518).
  • Cover the membrane completely during incubations to make sure it doesn’t dry out during the detection steps.
  • Make sure that the crosslinking is efficient.
  • Make sure that the 4X wash buffer is diluted to 1X.
  • Increase film exposure time during detection.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
With the Lightshift Chemiluminescent EMSA assay, how do I prevent all the DNA from shifting to the top of the gel?
Use a non-specific competitor DNA such as poly(dI.dC).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What causes speckling/spots on the blot in a Lightshift Chemiluminescent EMSA assay?
Speckling/spots can be caused by precipitate in the HRP conjugate or by air bubbles. Precipitate in the HRP conjugate can be removed by filtering the conjugate through a 0.2 µm filter or by centrifugation for 1 minute at maximum speed while air bubbles between the gel and the membrane should be removed before transfer.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I see very high background in my probed blot when using the Lightshift Chemiluminescent EMSA Kit. What could be the cause of this?
Here are possible causes and solutions:
- Particulate in Blocking Buffer or Wash Buffer: Gently warm until no particulate remains.
- Contaminants in the TBE: Use high-quality reagents or filter TBE through a 0.2 µm filter before use.
- The transfer unit or sponges used were dirty: Use clean equipment and sponges that were not previously used for western blotting.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I use NativePAGE gels to perform EMSA (electrophoretic mobility shift assays)?
NativePAGE gels have been successfully used to perform EMSA showing protein-protein interactions between two purified proteins, but we have not tested NativePAGE gels for EMSA analysis of interactions between nucleic acids (either DNA or RNA) and a protein or protein complex.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.