Trypsin, TPCK Treated - FAQs
Trypsin, TPCK Treated - FAQs
View additional product information for Trypsin, TPCK Treated - FAQs (20233)
1 product FAQs found
Do you have a protocol for making stock solution of Trypsin, TPCK Treated (Cat. No. 20233)?
This product should primarily be used as a powder directly in the digestion buffer.
Materials:
Digestion Buffer
0.1 M Ammonium carbonate (NH4)HCO3, pH 8.0.
Procedure:
1. Dissolve approximately 10 mg of the protein (=substrate) in 1 ml of Digestion Buffer.
2. Add sufficient TPCK Trypsin to the protein solution to have an enzyme: substrate ratio between 1:20 and 1:100 (w/w).
3. Incubate sample for 2–18 hours at 37 degrees C.
Note: The exact incubation time and enzyme: substrate ratio will have to be determined for each protein individually.
References:
Walsh, K.A., Neurath, H. (1964). Trypsinogen and chymotrypsinogen as homologous proteins. Proc. Natl. Acad. Sci. USA, 52, p. 884.
Cunningham, L.W. (1954). Molecular-kinetic properties of crystalline diisopropyl phosphoryl trypsin. J. Biol. Chem. 211, p.13.
Kostka, V., Carpenter, F. H. (1964). Inhibition of chymotrypsin activity in crystalline trypsin preparations. J. Biol. Chem. 239, 1799–1803.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.