beta-Methyl-DL-phenylalanine hydrochloride, 99%
beta-Methyl-DL-phenylalanine hydrochloride, 99%
beta-Methyl-DL-phenylalanine hydrochloride, 99%
Thermo Scientific Chemicals

beta-Methyl-DL-phenylalanine hydrochloride, 99%

CAS: 80997-87-1 | C10H14ClNO2 | 215.68 g/mol
Have Questions?
Catalog NumberQuantity
2026600101 g
Catalog number 202660010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS80997-87-1
IUPAC Name2-amino-3-phenylbutanoic acid hydrochloride
Molecular FormulaC10H14ClNO2
InChI KeySGKWQBMDOXGYAA-UHFFFAOYNA-N
SMILESCl.CC(C(N)C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Titration with HClO4>=98.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only