Immobilized Papain (Agarose Resin) - FAQs
Immobilized Papain (Agarose Resin) - FAQs
View additional product information for Immobilized Papain (Agarose Resin) - FAQs (20341)
2 product FAQs found
Can I purchase the immobilized enzymes (proteases) from the Thermo Scientific IgG Fragmentation kit separately?
Yes. The three immobilized enzymes are available in 5 mL package sizes as shown below:
• Immobilized Ficin, Cat. No. 44881
• Immobilized Papain, Cat. No. 20341
• Immobilized Pepsin, Cat. No. 20343
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How much of the immobilized Papain (Agarose Resin) do I need for digestion of 10 mg IgG?
0.5 mL of the 50% Immobilized Papain slurry is sufficient for the digestion of up to 10 mg of pure lyophilized IgG reconstituted in 1 mL of Digestion Buffer.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.