Pierce™ Streptavidin Agarose, 5 mL - FAQs
Pierce™ Streptavidin Agarose, 5 mL - FAQs
View additional product information for Pierce™ Streptavidin Agarose - FAQs (20353, 20349, 20347)
5 product FAQs found
What is the particle size of the Pierce Streptavidin Agarose beads (Cat. No. 20347, 20353, 20349)?
The average particle size for the Pierce Streptavidin Agarose beads is 45 to 165 µm.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the recommend/ideal antigen:antibody ratio when using the Pierce Streptavidin Agarose (Cat. No. 20347) for doing an immunoprecipitation (with pre-incubating antigen and antibody)?
The antigen:antibody ratio will need to be determined empirically since one cannot be sure of the amount of specific antibody against the antigen present in the plasma. The starting point is the binding capacity of the High Capacity Streptavidin Agarose Resin (Cat. No. 20347) which is 1-3 mg of biotinylated BSA/mL of settled resin. The binding capacity will vary depending on the molecular weight of the biotinylated antigen. Let's assume the molecular weight of the antigen is close to that of BSA (66.4 kDa), you can use an average of 2 mg antibody or antigen/mL of resin to do the calculation. Determine how much resin they want to use and calculate how much biotinylated protein is needed and then empirically determine how much plasma is needed.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the shelf life for Pierce Streptavidin Agarose?
We guarantee the functionality of the product for one year from the date of shipment, if stored as recommended.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How many biotin binding-sites are there per streptavidin molecule?
Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the binding capacity of streptavidin agarose?
The binding capacity of streptavidin agarose is 15-20 µg (18-24 nmoles) of fluorescein biotin per mL of sedimented gel.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.