Pierce™ Protein G Agarose
Thermo Scientific™

Pierce™ Protein G Agarose

The Thermo Scientific NAb Protein G Spin Kits are convenient for rapid, small-scale affinity purification of antibodies from a varietyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2039725 mL
203982 mL
2039910 mL
Catalog number 20397
Price (USD)
1,996.65
Online exclusive
2,295.00
Save 298.35
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
1,996.65
Online exclusive
2,295.00
Save 298.35
Each
The Thermo Scientific NAb Protein G Spin Kits are convenient for rapid, small-scale affinity purification of antibodies from a variety of sample types. Each pre-filled microcentrifuge spin column of the immobilized protein resin enables quick purification of 11-15 mg of human IgG from 0.5-2 mL of serum or other sample. Included in each kit are sufficient collection tubes, buffers and a streamlined protocol for purifying at least two antibody samples.

Pierce Protein G Agarose is a premium-quality affinity resin for antibody purification. It consists of recombinant Protein G that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose (CL-6B). This particular variety of the affinity resin provides the most versatile combination of chromatographic features for high yield and high-purity purification of whole IgG from mammalian serum samples. The agarose beads have physical and chemical properties suitable for many affinity purification systems.

Features of Pierce Protein G Agarose:

• Protein G – immobilized Protein G is ideal for polyclonal IgG purification from mouse, human, cow, goat and sheep serum, including human IgG3 and mouse IgG1 isotypes
• Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
• Inert and stable – superior manufacturing method immobilizes Protein G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
• Standard capacity – Pierce Protein G Agarose has a normal load of immobilized Protein G, providing a binding capacity of 11 to 15 mg human IgG/mL resin

Protein G is a bacterial cell wall protein original from group G Streptococcus and now produced as a recombinant in E. coli. Like Protein A, Protein G binds to most mammalian immunoglobulins primarily through their Fc regions. Native Protein G contains two immunoglobulin binding sites, as well as albumin and cell surface binding sites. In the recombinant form of Protein G, these albumin and cell surface binding sites have been eliminated to reduce nonspecific binding when purifying immunoglobulins. Recombinant Protein G has a mass of approximately 21.6kDa, but its apparent size by SDS-PAGE is 31 to 34kDa. IgG-binding function is optimal at pH 5 but also occurs efficiently in near-neutral conditions (pH 7.0 to 7.2).

Compared to Protein A, Protein G binds a broader spectrum of IgG subclasses from human, mouse and rat serum. In particular, besides binding other isotypes just as well as Protein A, Protein G exhibits stronger binding to human IgG3, mouse IgG1 and all three isotypes of Rat IgG. Thus, Protein G is generally recommended for applications involving these species and isotypes of antibody. With regard to other species, Protein G provides higher-capacity binding for IgG from cow, goat and sheep, but it binds more weakly than Protein A to pig, guinea pig, dog and cat IgG.

Pierce Protein G Agarose is prepared using Thermo Scientific AminoLink Coupling Chemistry, which provides several advantages compared to traditional methods of ligand immobilization. AminoLink Immobilization results in conjugation between sugar monomers of the agarose beads and native lysine residues on the Protein A via simple amide bonds. Unlike typical cyanogen bromide (CNBr) immobilization, the AminoLink Method does not introduce any novel chemical groups that could cause undesired nonspecific binding and produces a stable, essentially irreversible bond. The result is a high-binding-capacity resin that retains functional immobilized Protein G through multiple rounds of antibody purification.

Pierce Protein G Agarose is effective for affinity purification of IgG from serum and other fluids of many mammalian species. Protein G is especially well suited for use with mouse antibodies (including IgG1) in addition to most IgG isotypes from human, goat and sheep samples.

Properties of crosslinked 6% beaded agarose (CL-6B):
• Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
• Average Particle Size: 45 to 165 microns
• Exclusion Limit: 10,000 to 4,000,000 daltons
• Maximum Volumetric Flow Rate: approx. 1 mL/minute (for 1 cm diameter column)
• Maximum Linear Velocity: 30 cm per hour
• Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

More Product Data
• Using antibody-binding proteins for antibody purification

Related Products
Pierce™ Protein G Agarose
NAb™ Protein G Spin Columns, 1 mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Purification TargetIgG Antibodies
Quantity25 mL
Column TypeAgarose Resin, Affinity
FormatBottle
Product LinePierce
Product TypeAgarose
Stationary PhaseProtein G
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C. Product is shipped at ambient temperature.