L(+)-Cysteine hydrochloride monohydrate, 98.5-101.5% (dry basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Cysteine hydrochloride monohydrate, 98.5-101.5% (dry basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, BP, and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2071-061 kg
2071-05100 g
Catalog number 2071-06
Price (USD)
-
Quantity:
1 kg
This product is GMP Manufactured and meets USP, BP, and EP specifications.
This product is GMP Manufactured and meets USP, BP, and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS4-6-7048
Chemical Name or MaterialL(+)-Cysteine hydrochloride monohydrate
ColorWhite
Melting Point168°C to 170°C
Molecular FormulaHSCH2CH(NH2)COOH·HCl·H2O
PackagingPoly bottle, tamper-evident cap
Synonym(R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, (R)-(+)-Cysteine hydrochloride monohydrate, Cysteine Hydrochloride monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, monohydrate
GradeMulti-Compendial/USP
Molecular Weight (g/mol)175.64
Percent Purity98.5 to 101.5%
Quantity1 kg
pH0.8 to 1.2 (100 g/L, H2O, 20°C)
Unit Size1 kg