L-Phenylalanine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L-Phenylalanine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.Más información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2086-061 kg
2086-0712 kg
Número de catálogo 2086-06
Precio (MXN)
-
Cantidad:
1 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición313°C (1013 hPa)
CAS63-91-2
Nombre del producto químico o materialL-Phenylalanine
ColorWhite
Punto de fusión173°C
Fórmula molecularC9H11NO2
EnvasePoly bottle, tamper-evident cap
Sinónimo(S)-(-)-Phenylalanine, H-L-Phe-OH, Phenylalanine, H-Phe-OH, 3-Phenyl-L-alanine
GradoMulticomponente/USP
Peso molecular (g/mol)165.19
Porcentaje de pureza98.5 to 101.5%
Cantidad1 kg
pH5.4 to 6
Unit SizeEach