MBP Column Preparation Buffer for Pierce™ IgM Purification Kit
MBP Column Preparation Buffer, formulation: Tris buffer with sodium chloride, sodium azide and EDTA; pH 7.4. For use with PierceRead more
Have Questions?
Catalog Number
Quantity
21018
50 mL
Catalog number 21018
Price (EUR)
62,34
Online exclusive
68,50
Save 6,16 (9%)
Each
Quantity:
50 mL
Price (EUR)
62,34
Online exclusive
68,50
Save 6,16 (9%)
Each
MBP Column Preparation Buffer, formulation: Tris buffer with sodium chloride, sodium azide and EDTA; pH 7.4. For use with Pierce Mannan Binding Protein Agarose, and the Pierce IgM Purification Kit.