Thermo Scientific Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on자세히 알아보기
Have Questions?
카탈로그 번호
수량
22212
10 mL
카탈로그 번호 22212
제품 가격(KRW)
2,021,000
온라인 행사
Ends: 30-Sep-2026
2,377,000
할인액 356,000 (15%)
Each
수량:
10 mL
제품 가격(KRW)
2,021,000
온라인 행사
Ends: 30-Sep-2026
2,377,000
할인액 356,000 (15%)
Each
Thermo Scientific Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5 mL column of the MBP Agarose.
Mannan binding protein (MBP), also known as mannose-binding lectin (MBL) is a mammalian lectin that is produced in the liver and occurs in serum. MBP consists of 18 identical subunits, each with a molecular mass of approximately 31kDa. The protein is capable of initiating carbohydrate-mediated complement activation.