Restore™ Western Blot Stripping Buffer
Thermo Scientific™

Restore™ Western Blot Stripping Buffer

Thermo Scientific Restore Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21059500 mL
21059X44 x 500 mL
2106230 mL
210635 L
Catalog number 21059
Price (USD)
181.65
Online exclusive
216.00
Save 34.35
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
181.65
Online exclusive
216.00
Save 34.35
Each

Thermo Scientific Restore Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.

Features of Restore Western Blot Stripping Buffer:

  • Saves time—no need to re-run gels and blots
  • Saves costly sample—re-probe the membrane using the same target sample
  • Effective—formulation is more efficient at stripping antibodies than homemade buffers
  • Gentle—does not damage the target antigen during stripping allowing efficient reprobing
  • Odor-free—no mercaptans means no acrid odors
  • Economical—less expensive than other commercial stripping buffers

Product Details

Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore Western Blot Stripping Buffer eliminates this waste when detecting immunoblots with chemiluminescent Western blotting substrates. Restore Stripping Buffer provides clean and efficient removal of primary and secondary antibodies from immunoblots without removing or damaging the immobilized antigen allowing blots to be stripped and reprobed with confidence.

Chemiluminescent Western blot detection with reagents such as Thermo Scientific SuperSignal Substrates for horseradish peroxidase is one of the most common and sensitive methods in use today. Because these substrates do not precipitate and bind to membrane surfaces, Western blots detected by chemiluminescence can be stripped with reagents that remove affinity-bound primary and secondary antibodies. To be effective, a stripping buffer must be strong enough to disassociate bound antibodies but gentle enough to leave the transferred target proteins intact on the nitrocellulose or PVDF membrane. Restore Western Blot Stripping Buffer has these characteristics.

By stripping and reprobing, there is no need to waste rare or costly samples by running multiple gels in order to probe for different targets. A single membrane from one gel can be stripped with Restore Western Blot Stripping Buffer to remove the primary antibodies. Stripping the blot takes only 15 to 30 minutes, depending on the affinity of the primary antibody. After stripping, block and reprobe with a new primary antibody. Alternatively, a blot can be stripped and reprobed with adjusted antibody concentrations to optimize conditions after obtaining initially poor results.

Applications

  • Reuse a nitrocellulose or PVDF blot to detect a different target with a different primary antibody
  • Reprobe a blot to correct or optimize antibody concentrations that were ineffective the first time

Protocol Summary

  • Wash blot to remove chemiluminescent substrate.
  • Incubate blot in Restore Western Blot Stripping Buffer for 5 to 15 minutes at 37°C (room temperature is sufficient for some antibodies).
  • Remove blot and wash in Wash Buffer (TBS-T or PBS-T).
  • Test for sufficient removal of antibodies.
  • Perform next immunoblot experiment.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LineRestore
Product TypeWestern Blot Stripping Buffer
Quantity500 mL
Unit SizeEach
Contents & Storage
Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.

Frequently asked questions (FAQs)

How many times can I strip my blot using Restore or Restore Plus Western Blot Stripping Buffers ?

The stability of the attached (transferred and bound) protein will determine the number of times the membrane can be successfully re-probed after stripping. The protein may withstand stripping as many as four times or as few as one time.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the proper wash buffer to use after stripping with Restore or Restore Plus Western Blot Stripping buffers?

Wash the membrane with the same buffer as was used between antibody probing steps during the Western blotting procedure.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I strip membranes with Restore or Restore Plus Western Blot Stripping Buffer and reprobe for subsequent detection with fluorescent probes?

No. Although the fluorescent antibodies, like other antibodies, can be stripped with Restore Buffers, stripped membranes typically produce unacceptable background for subsequent fluorescent detection methods. We recommend Restore Fluorescent Western Blot Stripping Buffer, Cat. No. 62299 (20 mL) and Cat. No. 62300 (100 mL).

Note: Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane (Cat. No. 22860) only.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Will Restore and Restore Plus Western Blot Stripping Buffers remove biotin-bound avidin, streptavidin or NeutrAvidin Protein conjugates?

No. Restore and Restore Plus Buffers are not stringent enough to break the avidin-biotin interaction. A buffer capable of breaking this interaction would likely also damage the target protein, making it undetectable. However it may be possible to strip the complexed biotinylated primary antibody and streptavidin-HRP secondary antibody from the target protein on the membrane. Please also see Tech Tip: Strip and Reprobe Western Blots (https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/strip-reprobe-western-blots-tech-tip.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Will the Restore and Restore Plus Western Blot Stripping Buffers remove precipitating substrates?

No. The antibodies are removed but the substrate leaves a permanent precipitate on the membrane that cannot be removed. Restore and Restore Plus Buffers are designed for procedures using chemiluminescent substrates. Please note that this is not compatible with fluorescence supstrates as it will result in increased background. For fluorescent substrates please use our Restore Fluorescent Western Blot Stripping Buffer. Please also see Tech Tip: Strip and Reprobe Western Blots (https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/strip-reprobe-western-blots-tech-tip.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.