Nalgene™ Square Wide-Mouth PPCO Bottles with Closure, 500mL - FAQs
View additional product information for Nalgene™ Square Wide-Mouth PPCO Bottles with Closure - FAQs (2110-0002, 2110-0032PK, 2110-0006, 2110-0008PK, 2110-0016, 2110-0008, 2110-0006PK, 2110-0016PK, 2110-0002PK, 2110-0032)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.