Pierce™ Streptavidin, Maleimide-Activated - FAQs
View additional product information for Pierce™ Streptavidin, Maleimide-Activated - FAQs (21102)
1 product FAQs found
What is the molecular weight of Pierce Streptavidin, Maleimide-Activated (Cat. No. 21102)?
The molecular weight is 60 kDa.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.