Can I strip and reprobe my blot after using the Clean-Blot IP Detection reagent?
Yes. When using a chemiluminescent substrate, membranes can be stripped and reprobed similar to probing with secondary antibodies.