Nalgene™ Fluorinated Wide-Mouth HDPE Bottles with Closure, 125mL - FAQs
Nalgene™ Fluorinated Wide-Mouth HDPE Bottles with Closure, 125mL - FAQs
View additional product information for Nalgene™ Fluorinated Wide-Mouth HDPE Bottles with Closure - FAQs (2197-0008PK, 2197-0016, 2197-0004, 2197-0016PK, 2197-0032, 2197-0004PK, 2197-0008, 2197-0032PK)
1 product FAQs found
To what level are your FLPE fluorinated containers processed?
All of our fluorinated products are processed to Level 5.