Dionex™ Inuvion™ Core IC System - FAQs
Dionex™ Inuvion™ Core IC System - FAQs
View additional product information for Dionex™ Inuvion™ Core IC System - FAQs (22185-60100)
11 product FAQs found
Can the Dionex Inuvion Core IC system be upgraded?
Yes. The Dionex Inuvion Core can be upgraded to the Dionex Inuvion IC system feature set, which includes electrolytic suppression and a vacuum degasser. Once upgraded to the Dionex Inuvion IC system, you can further upgrade to include RFIC eluent generation, providing a pathway to automation and reagent-free operation without replacing the base instrument.
What are the key features of the Dionex Inuvion Core IC system?
The Dionex Inuvion Core IC system is designed for labs that need dependable IC performance at a lower cost, while retaining the flexibility to upgrade to full RFIC functionality later. It includes a high-pressure pump, chemical suppression, and requires manual eluent preparation.
What accessories can I add to the Dionex Inuvion IC system?
We offer multiple options and peripherals to extend the capabilities of the Dionex Inuvion IC system. Available accessories include:
  • An additional 6-port or 10-port valve
  • Column heater
  • Digital gas pressure regulator
  • IC PEEK tubing with Viper fittings
  • An integrated regenerant pump
  • Seal wash pump
  • Eluent monitor (for 2 L or 4 L bottles)
You can easily swap these accessories in and out of the instrument in less than 5 minutes, allowing you to customize the system as your application needs change.
Can the Dionex Inuvion IC system be upgraded?
Yes. It’s a modular, upgradeable platform. You can start with the Dionex Inuvion Core IC system and later add electrolytic suppression or Reagent-Free IC (RFIC) capabilities as your analytical requirements expand. Also, you can easily swap out many of the accessories, such as auxiliary valves and the integrated regenerant pump if your application changes.
How is the reliability and performance of the Dionex Inuvion IC System?
The Dionex Inuvion IC system’s high-pressure pump enables precise, continuous flows at up to 5000 psi with vacuum degassing of the eluent. When combined with a precise temperature-controlled conductivity cell delivery, the system has a very stable baseline which produces excellent results. Additionally, automated eluent generation with monitoring and self-serviceable parts minimize downtime and maximize reliability.
What types of samples and analyses can be performed on the Dionex Inuvion IC?
It supports ion analysis in diverse matrices such as drinking water, wastewater, environmental samples, and industrial process streams. The system accommodates isocratic and gradient separations on a broad range of 2 mm and 4 mm columns for different ionic species.
What makes the Dionex Inuvion IC system easier to use than previous IC instruments?
The Dionex Inuvion platform is built with new, advanced electronics and a high-performance pump to improve the speed and quality of your results. It features a smart, function-driven design with easy access to all components, built-in how-to videos, automated startup/shutdown, and self-diagnostics. This minimizes training time and operator error while maximizing uptime.
What is the Thermo Scientific Dionex Inuvion Ion Chromatography system?
The Dionex Inuvion IC system is a next-generation ion chromatography platform designed for reliable, high-performance ion analysis. It combines intuitive operation, automated workflows, and flexible configurations for consistent, reproducible results every day—ideal for academic, municipal, analytical, and industrial labs performing routine or research IC applications.
What kind of support will I get for my Dionex ion chromatography system? 
 We provide comprehensive support. Our specialists act as trusted advisors and share industry expertise. They understand that purchasing an IC instrument is also purchasing a relationship, and they strive to provide the level of service and support that laboratories need. 
How does the Dionex Integrion HPIC system improve laboratory throughput?  
 By increasing pressure tolerance up to 6000 psi, smaller particle size columns such as 4 µm particle-size columns can be used to achieve shorter run times. This allows the system to provide increased throughput and productivity without sacrificing resolution or data quality. Automated eluent generation provides time savings by eliminating human errors associated with manual eluent preparation. 
What does HPIC stand for in Thermo Scientific Dionex ion chromatography systems and why is it important?  
 HPIC stands for high-pressure ion chromatography. This capability enables the use of smaller-particle-size 4 µm columns, which provide faster flow rates without sacrificing resolution. HPIC results in shorter run times and increased laboratory throughput while maintaining data quality.