Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle - FAQs
View additional product information for Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle - FAQs (2226-0020PK)
3 product FAQs found
What is the vacuum rating for Nalgene Heavy-Duty PPCO Vacuum Bottles with Closure?
These bottles can hold full vacuum for up to 24 hrs at 20 degrees C.
How strong are the handles of your Nalgene polypropylene carboys and how do you test the strength of the handles?
Nalgene polypropylene carboys are safe to lift by the handles when full, as they are capable of supporting at least double their intended weight on a single handle.
To test the strength of the carboy handles, carboys are suspended through a single handle on the carboy body. 10 L, 20 L, and 50 L carboys are tested by filling the container with steel ballast pellets to a weight in excess of double the weight of the carboy filled to nominal capacity with water.
I need a container with a positive pressure rating. Do you have anything suitable for this?
Unfortunately, none of our containers were designed or tested for applications involving positive pressure. Some of our customers do choose to carry out their own performance validations, but this must be done at the customer‘s own risk. We do offer selected bottles and carboys (2126 and 2226 series) that are vacuum rated for use at room temperature (to full “rough” vacuum, about 28-29 in. Hg).