L-Photo-Leucine
Thermo Scientific™

L-Photo-Leucine

Thermo Scientific Pierce L-Photo-Leucine is an analog of L-leucine that can be incorporated into proteins during synthesis and then light-activatedRead more
Have Questions?
Catalog NumberQuantity
22610100 mg
Catalog number 22610
Price (USD)
618.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
618.00
Each
Thermo Scientific Pierce L-Photo-Leucine is an analog of L-leucine that can be incorporated into proteins during synthesis and then light-activated to covalently crosslink interacting proteins in cells.

Features of L-Photo-Leucine:

• In vivo labeling—incorporate photo-reactive group into proteins using normal cellular machinery of live cells
• In vivo crosslinking—find interacting proteins in the native cellular environment
• Increased specificity—crosslink interacting proteins correctly positioned at their interfaces within protein interaction domains
• Efficient recovery—90% protein recovery in cell lysates after crosslinking
• Compatible—crosslink proteins expressed in a wide variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3 and C6
• Easy to use—reagents are photo-stable under normal laboratory lighting, eliminating the need to work in the dark

Thermo Scientific L-Photo-Leucine is an analog of L-leucine that can be endogenously incorporated into the primary sequence of proteins during synthesis and then ultraviolet light (UV)-activated to covalently crosslink proteins within protein-protein interaction domains in their native environment within live cells. The powerful method enables both stable and transient protein interactions in cells to be studied and characterized without the use of completely foreign chemical crosslinkers and associated reagent solvents during the interaction experiment that can adversely aspects of the cell biology being studied.

When used in combination with specially formulated limiting media that is devoid of leucine, the photo-activatable derivative is treated like a naturally occurring amino acid by the protein synthesis machinery within the cell. As a result, it can be substituted for leucine in the primary sequence of proteins during catabolism and growth. Photo-leucine derivatives contain diazirine rings that activate when exposed to UV light to become reactive intermediates that form covalent bonds with nearby protein side chains and backbones. Naturally associating binding partners within the cell can be instantly trapped by photoactivation of the diazirine-containing proteins in the cultured cells. Crosslinked protein complexes can be detected by decreased mobility on SDS-PAGE followed by western blot detection, size exclusion chromatography, sucrose density gradient sedimentation or mass spectrometry.

Resources:

Photoreactive Amino Acid FAQ

Related Products
L-Photo-Methionine
Dulbecco's Modified Eagle's Limiting Medium (DMEM-LM)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityYes
DescriptionL-Photo-Leucine
FormSolid
Labeling MethodChemical Labeling, Metabolic Labeling
Molecular Weight (g/mol)143.15
PEGylatedNo
Product LinePierce
Quantity100 mg
Reactive MoietyDiazirine
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length0.0 Å
Water SolubleYes
Chemical ReactivityNonselective-NA (Metabolic labeling)
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C protected from light.

Frequently asked questions (FAQs)

Apart from proteins, will the Photoreactive Amino Acids crosslink to other molecules?

Photoreactive amino acids are incorporated into proteins during synthesis; but upon UV activation, they can crosslink to other biomolecules within proximity.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How stable are the Photoreactive Amino Acids in DMEM-LM?

Long-term stability of the photoreactive amino acids in media has not been determined but should be comparable to their natural analogs, if protected from light. For best results, store the photoreactive amino acids at -20°C as a dry compound. Just before use, add the photoreactive amino acids to the minimal volume of DMEM-LM supplemented with dialyzed serum.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How long do I need to incubate the Photoreactive Amino Acids with my cells?

Incubate the photoreactive amino acids in DMEM-LM with cells for 24 hours. For proteins with high turnover, a minimum incubation of 8 hours is needed for detectable crosslinking levels. Incubation for longer than 24 hours might significantly affect cell growth or viability.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

My cells will not grow in DMEM, what other type of culture media can be used with the Photoreactive Amino Acids?

Some cells types can be adapted to grow in DMEM before using the DMEM-LM supplemented with the photoreactive amino acids. Currently we do not offer any other leucine- and methionine-depleted culture medium.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I use the Photoreactive Amino Acids to crosslink peptides?

Peptides can be synthesized using the N-termini-protected (Boc or Fmoc) photoreactive amino acid derivatives.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (3)

Citations & References
Abstract
Mass spectrometry of laser-initiated carbene reactions for protein topographic analysis.
Authors:Jumper CC, Schriemer DC
Journal:Anal Chem
PubMed ID:21425771
'We report a protein labeling method using nonselective carbene reactions of sufficiently high efficiency to permit detection by mass spectrometric methods. The approach uses a diazirine-modified amino acid (l-2-amino-4,4''-azipentanoic acid, "photoleucine") as a label source, which is converted to a highly reactive carbene by pulsed laser photolysis at 355 nm. ... More
The copper binding domain of SPARC mediates cell survival in vitro via interaction with integrin beta1 and activation of integrin-linked kinase.
Authors:Weaver MS, Workman G, Sage EH
Journal:J Biol Chem
PubMed ID:18503049
'Secreted protein acidic and rich in cysteine (SPARC) is important for the normal growth and maintenance of the murine lens. SPARC-null animals develop cataracts associated with a derangement of the lens capsule basement membrane and alterations in lens fiber morphology. Cellular stress and disregulation of apoptotic pathways within lens epithelial ... More
Inhibition of calcineurin-mediated endocytosis and alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) receptors prevents amyloid beta oligomer-induced synaptic disruption.
Authors:Zhao WQ, Santini F, Breese R, Ross D, Zhang XD, Stone DJ, Ferrer M, Townsend M, Wolfe AL, Seager MA, Kinney GG, Shughrue PJ, Ray WJ
Journal:J Biol Chem
PubMed ID:20032460
Synaptic degeneration, including impairment of synaptic plasticity and loss of synapses, is an important feature of Alzheimer disease pathogenesis. Increasing evidence suggests that these degenerative synaptic changes are associated with an accumulation of soluble oligomeric assemblies of amyloid beta (Abeta) known as ADDLs. In primary hippocampal cultures ADDLs bind to ... More