6-Chloropurine riboside, 99+%
6-Chloropurine riboside, 99+%
6-Chloropurine riboside, 99+%
Thermo Scientific Chemicals

6-Chloropurine riboside, 99+%

CAS: 5399-87-1 | C10H11ClN4O4 | 286.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226450010
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Chemical Identifiers
CAS5399-87-1
IUPAC Name2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC10H11ClN4O4
InChI KeyXHRJGHCQQPETRH-UHFFFAOYNA-N
SMILESOCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only