2'-Deoxyuridine, 99+%
2'-Deoxyuridine, 99+%
2'-Deoxyuridine, 99+%
Thermo Scientific Chemicals

2'-Deoxyuridine, 99+%

CAS: 951-78-0 | C9H12N2O5 | 228.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226612500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS951-78-0
IUPAC Name1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H12N2O5
InChI KeyMXHRCPNRJAMMIM-SHYZEUOFSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration with NaOH>=99.0 % (On dry substance)
HPLC>=99.0 %
Heavy metals (as Pb)=<10 ppm
Loss on drying=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only